2-Phthalimidopropionic acid 98%
Lieferant: Apollo Scientific
OR6527-25G
OR6527-100G
APOSOR6527-25GEA
45.9
EUR
APOSOR6527-25G
APOSOR6527-100G
2-Phthalimidopropionic acid 98%
2-Phthalimidopropionic acid
Formel:
C₁₁H₉NO₄ Schmelzpunkt: 159…164 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00023061 CAS-Nummer: 19506-87-7 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...