4-(Hydroxymethyl)cyclohexanon 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR6431-1G
OR6431-5G
APOSOR6431-1GEA
171
EUR
APOSOR6431-1G
APOSOR6431-5G
4-(Hydroxymethyl)cyclohexanon 95%
4-(Hydroxymethyl)cyclohexanone
Formel:
C₇H₁₂O₂ Siedepunkt: 105…110 °C/0.08mmHg Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08056334 CAS-Nummer: 38580-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...