4-Oxocyclohexanecarbonyl chloride
Lieferant: Apollo Scientific
OR6382-1G
OR6382-250MG
OR6382-500MG
APOSOR6382-1GEA
0
EUR
APOSOR6382-1G
APOSOR6382-250MG
APOSOR6382-500MG
4-Oxocyclohexanecarbonyl chloride
4-Oxocyclohexanecarbonyl chloride
Formel:
C₇H₉ClO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08056325 CAS-Nummer: 914637-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...