4-(1H-Imidazol-4-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6239-1G OR6239-250MG OR6239-100MG
APOSOR6239-1GEA 0 EUR
APOSOR6239-1G APOSOR6239-250MG APOSOR6239-100MG
4-(1H-Imidazol-4-yl)benzoic acid 97%
4-(1H-Imidazol-4-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Schmelzpunkt: 308 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08056298
CAS-Nummer: 13569-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR