5-Dihydroxyboronylnicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR6227-1G
OR6227-250MG
APOSOR6227-1GEA
561
EUR
APOSOR6227-1G
APOSOR6227-250MG
5-Dihydroxyboronylnicotinic acid
5-Dihydroxyboronylnicotinic acid
Formel:
C₆H₆BNO₄ Schmelzpunkt: 226…230 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02323212 CAS-Nummer: 913836-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...