2-Methyl-2-propanyl-(2-hydroxyethyl)methylcarbamat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(tert-Butoxycarbonyl)-N-methylethanolamine, N-Boc-N-methyl-2-aminoethanol, tert-Butyl (2-Hydroxyethyl)(methyl)carbamate, N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol, N-Boc-N-methylethanolamine, N-Boc-N-methylglycinol, N-(tert-Butoxycarbonyl)-N-methylglycinol, (2-Hydroxyethyl)(methyl)carbamic acid tert-Butyl ester
OR61447-10G
OR61447-100G
OR61447-25G
OR61447-5G
APOSOR61447-10GEA
94.4
EUR
APOSOR61447-10G
APOSOR61447-100G
APOSOR61447-25G
APOSOR61447-5G
2-Methyl-2-propanyl-(2-hydroxyethyl)methylcarbamat 97%
tert-Butyl (2-hydroxyethyl)(methyl)carbamate
Formel:
C₈H₁₇NO₃ Siedepunkt: 83…84 °C/0.05mmHg Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD03425857 CAS-Nummer: 57561-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...