2-(4-Formyl-3-methoxyphenoxy)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Formyl-3-methoxy-phenoxy)-acetic acid
OR61394-1G
OR61394-5G
APOSOR61394-1GEA
179
EUR
APOSOR61394-1G
APOSOR61394-5G
2-(4-Formyl-3-methoxyphenoxy)acetic acid
2-(4-Formyl-3-methoxyphenoxy)acetic acid
Formel:
C₁₀H₁₀O₅ Schmelzpunkt: 207…209 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058468 CAS-Nummer: 84969-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...