L-Thr(Me)-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: O-Methyl-L-Threonine, H-Thr(Me)-OH, (2S,3R)-2-Amino-3-methoxybutanoic acid

OR61331-1G OR61331-5G OR61331-250MG
APOSOR61331-1GEA 39.3 EUR
APOSOR61331-1G APOSOR61331-5G APOSOR61331-250MG
L-Thr(Me)-OH
L-Thr(Me)-OH
Formel: C₅H₁₁NO₃
Schmelzpunkt: 213…215 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00037767
CAS-Nummer: 4144-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR