Piperidin-3-amine dihydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

OR6131-10G OR6131-25G OR6131-5G
APOSOR6131-10GEA 136 EUR
APOSOR6131-10G APOSOR6131-25G APOSOR6131-5G
Piperidin-3-amine dihydrochloride 95+%
Piperidin-3-amine dihydrochloride
Formel: C₅H₁₄Cl₂N₂
Schmelzpunkt: 230 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012773
CAS-Nummer: 138060-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR