(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Lys(Z)-OMe.HCl, H-Lys(Z)-OMe·HCl
OR60214-100G
OR60214-25G
APOSOR60214-100GEA
328
EUR
APOSOR60214-100G
APOSOR60214-25G
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Formel:
C₁₅H₂₃ClN₂O₄ Schmelzpunkt: 115…118 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00034846 CAS-Nummer: 27894-50-4 EINECS: 248-715-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...