3-Brom-2-cyan-5-methylpyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-methylpicolinonitrile, 3-Bromo-5-methylpyridine-2-carbonitrile
OR60144-1G
OR60144-10G
OR60144-5G
APOSOR60144-1GEA
87.6
EUR
APOSOR60144-1G
APOSOR60144-10G
APOSOR60144-5G
3-Brom-2-cyan-5-methylpyridin 95%
3-Bromo-2-cyano-5-methylpyridine
Formel:
C₇H₅BrN₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD16988815 CAS-Nummer: 474824-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...