6-Amino-2,4-dimethylphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4,6-dimethylphenol
OR60001-1G
OR60001-5G
APOSOR60001-1GEA
42.2
EUR
APOSOR60001-1G
APOSOR60001-5G
6-Amino-2,4-dimethylphenol 98%
6-Amino-2,4-xylenol
Formel:
C₈H₁₁NO Schmelzpunkt: 130 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007691 CAS-Nummer: 41458-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...