3,4-Dihydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Protocatechuic aldehyde, Protocatechualdehyde

OR5940-100G OR5940-25G OR5940-10G OR5940-500G
APOSOR5940-100GEA 30.6 EUR
APOSOR5940-100G APOSOR5940-25G APOSOR5940-10G APOSOR5940-500G
3,4-Dihydroxybenzaldehyd
3,4-Dihydroxybenzaldehyd
Formel: C₇H₆O₃
Schmelzpunkt: 150…157 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003370
CAS-Nummer: 139-85-5
EINECS: 205-377-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR