4-Brombenzamidin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

OR59369-1G OR59369-5G
APOSOR59369-1GEA 52.2 EUR
APOSOR59369-1G APOSOR59369-5G
4-Brombenzamidin Hydrochlorid 98%
4-Brombenzamidin Hydrochlorid
Formel: C₇H₈BrClN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00464967
CAS-Nummer: 55368-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR