N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
N,N-Dimethyl-1-piperidin-3-ylmethanamine
OR5905-1G
OR5905-500MG
OR5905-5G
APOSOR5905-1GEA
401
EUR
APOSOR5905-1G
APOSOR5905-500MG
APOSOR5905-5G
N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride 95%
N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride
Formel:
C₈H₁₈N₂·2HCl Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13561484 CAS-Nummer: 90203-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...