2'-(Methylsulfonyl)acetanilid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Acetyl-2-(methylsulphonyl)aniline
OR5729-1G
OR5729-25G
OR5729-5G
APOSOR5729-1GEA
21.9
EUR
APOSOR5729-1G
APOSOR5729-25G
APOSOR5729-5G
2'-(Methylsulfonyl)acetanilid
2'-(Methylsulfonyl)acetanilid
Formel:
C₉H₁₁NO₃S Schmelzpunkt: 143.5…147.5 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01464107 CAS-Nummer: 20628-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...