2-Morpholinoacetic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Morpholinoacetic acid hydrochloride, Morpholin-4-ylacetic acid hydrochloride, 4-Morpholineacetic acid hydrochloride
OR5654-1G
OR5654-10G
OR5654-5G
APOSOR5654-1GEA
21.9
EUR
APOSOR5654-1G
APOSOR5654-10G
APOSOR5654-5G
2-Morpholinoacetic acid hydrochloride
2-Morpholinoacetic acid hydrochloride
Formel:
C₆H₁₂ClNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03540888 CAS-Nummer: 89531-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...