2-Boc-aminothiazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-1,3-thiazole-4-carboxylic acid, 2-BOC protected, 2-Boc-Aminothiazole-4-carboxylic acid

OR5596-1G OR5596-5G
APOSOR5596-1GEA 21.9 EUR
APOSOR5596-1G APOSOR5596-5G
2-Boc-aminothiazole-4-carboxylic acid 95%
2-Boc-aminothiazole-4-carboxylic acid
Formel: C₉H₁₂N₂O₄S
Schmelzpunkt: 250…252 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02181057
CAS-Nummer: 83673-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR