2-Methyl-2-(pyrazin-2-yl)propan-1-amine 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(1-Amino-2-methylprop-2-yl)pyrazine

OR55001-1G OR55001-5G
APOSOR55001-1GEA 592 EUR
APOSOR55001-1G APOSOR55001-5G
2-Methyl-2-(pyrazin-2-yl)propan-1-amine 95+%
2-Methyl-2-(pyrazin-2-yl)propan-1-amine
Formel: C₈H₁₃N₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11898940
CAS-Nummer: 1176768-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR