2-(Benzimidazolylthio)acetic acid hydrazide 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-((1H-Benzo[d]imidazol-2-yl)thio)acetohydrazide
OR5453-25G
OR5453-5G
APOSOR5453-25GEA
343
EUR
APOSOR5453-25G
APOSOR5453-5G
2-(Benzimidazolylthio)acetic acid hydrazide 97%
2-(Benzimidazolylthio)acetic acid hydrazide
Formel:
C₉H₁₀N₄O₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00171027 CAS-Nummer: 30065-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...