2,4-Diaminophenol Dihydrochlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

OR53100-100G OR53100-25G
APOSOR53100-100GEA 139 EUR
APOSOR53100-100G APOSOR53100-25G
2,4-Diaminophenol Dihydrochlorid
2,4-Diaminophenol Dihydrochlorid
Formel: C₆H₁₀Cl₂N₂O
Schmelzpunkt: 222 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012979
CAS-Nummer: 137-09-7
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR