Methyl-2-methylquinoline-6-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-methylquinoline-6-carboxylate
OR52927-25G
OR52927-5G
APOSOR52927-25GEA
313
EUR
APOSOR52927-25G
APOSOR52927-5G
Methyl-2-methylquinoline-6-carboxylate 98%
Methyl-2-methylquinoline-6-carboxylate
Formel:
C₁₂H₁₁NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02690144 CAS-Nummer: 108166-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...