Ethylvanillat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-hydroxy-3-methoxybenzoate

OR5260-100G OR5260-1KG OR5260-500G
APOSOR5260-100GEA 40.6 EUR
APOSOR5260-100G APOSOR5260-1KG APOSOR5260-500G
Ethylvanillat 97%
Ethylvanillat
Formel: C₁₀H₁₂O₄
Schmelzpunkt: 39…44 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00017269
CAS-Nummer: 617-05-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR