L(+)-Lysin Monohydrat 98%

Lieferant: Apollo Scientific

Synonyme: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate

OR52570-100G OR52570-500G OR52570-5G OR52570-1KG
APOSOR52570-100GEA 72.5 EUR
APOSOR52570-100G APOSOR52570-500G APOSOR52570-5G APOSOR52570-1KG
L(+)-Lysin Monohydrat 98%
L(+)-Lysin Monohydrat
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
Molekulargewicht: 164.2 g/mol
Schmelzpunkt: 212…214 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151035
CAS-Nummer: 39665-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR