H-D-Ala-OiPr·HCl
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
D-Alanine isopropyl ester hydrochloride, H-D-Ala-OiPr.HCl
OR52564-100G
OR52564-25G
OR52564-5G
APOSOR52564-100GEA
241
EUR
APOSOR52564-100G
APOSOR52564-25G
APOSOR52564-5G
H-D-Ala-OiPr·HCl
H-D-Ala-OiPr·HCl
Formel:
C₆H₁₃NO₂·HCl Schmelzpunkt: 85 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD17976853 CAS-Nummer: 39613-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...