3-Brom-4-nitroanilin 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR5232-25G
OR5232-5G
OR5232-100G
APOSOR5232-25GEA
62.5
EUR
APOSOR5232-25G
APOSOR5232-5G
APOSOR5232-100G
3-Brom-4-nitroanilin 99%
3-Brom-4-nitroanilin
Formel:
C₆H₅BrN₂O₂ Schmelzpunkt: 174…177 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07783047 CAS-Nummer: 40787-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...