Bis(9,9-dimethyl-9H-fluoren-2-yl)amine 99%

Lieferant: Apollo Scientific

OR51011-1G OR51011-5G
APOSOR51011-1GEA 0 EUR
APOSOR51011-1G APOSOR51011-5G
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine 99%
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine
Formel: C₃₀H₂₇N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD23135317
CAS-Nummer: 500717-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR