3,5-Dimethylbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR4988-100G
OR4988-1KG
OR4988-500G
APOSOR4988-100GEA
21.9
EUR
APOSOR4988-100G
APOSOR4988-1KG
APOSOR4988-500G
3,5-Dimethylbenzoesäure 98%
3,5-Dimethylbenzoesäure
Formel:
C₉H₁₀O₂ Schmelzpunkt: 169…171 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002525 CAS-Nummer: 499-06-9 EINECS: 207-876-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...