Terephthalaldehyde mono(diethyl acetal) 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Diethoxymethyl)benzaldehyde

OR4985-100G OR4985-25G
APOSOR4985-100GEA 263 EUR
APOSOR4985-100G APOSOR4985-25G
Terephthalaldehyde mono(diethyl acetal) 97%
Terephthalaldehyde mono(diethyl acetal)
Formel: C₁₂H₁₆O₃
Siedepunkt: 89…93 °C/7mmHg
Dichte: 1.047 g/cm³ (25 °C)
Flammpunkt: >113 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010217
CAS-Nummer: 81172-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR