α-Brom-2,5-dichlortoluol

Lieferant: Apollo Scientific

Synonyme: 2,5-Dichlorobenzyl bromide, 2-(Bromomethyl)-1,4-dichlorobenzene

OR49015-25G OR49015-100G
APOSOR49015-25GEA 119 EUR
APOSOR49015-25G APOSOR49015-100G
α-Brom-2,5-dichlortoluol
α-Brom-2,5-dichlortoluol
Formel: C₇H₅BrCl₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00236034
CAS-Nummer: 85482-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR