6-Bromo-3-chloro-2-cyanopyridine 95%
Lieferant: Apollo Scientific
Synonyme:
6-Bromo-3-chloropicolinonitrile
OR480829-1G
OR480829-5G
APOSOR480829-1GEA
145
EUR
APOSOR480829-1G
APOSOR480829-5G
6-Bromo-3-chloro-2-cyanopyridine 95%
6-Bromo-3-chloro-2-cyanopyridine
Formel:
C₆H₂BrClN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD17014673 CAS-Nummer: 1252046-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...