Boc-L-valine hydroxysuccinimide ester 97%
Lieferant: Apollo Scientific
Synonyme: N-Boc-L-valine N-succinimidyl ester, Boc-Val-OSu
OR480668-100G OR480668-5G OR480668-25G
APOSOR480668-100GEA 190 EUR
APOSOR480668-100G APOSOR480668-5G APOSOR480668-25G
Boc-L-valine hydroxysuccinimide ester 97%
Boc-L-valine hydroxysuccinimide ester
Formel: C₁₄H₂₂N₂O₆
Molekulargewicht: 314.338 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00037906
CAS-Nummer: 3392-12-9