L-Prolinbenzylester -hydrochlorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Pro-OBzl.HCl
OR480596-100G
OR480596-25G
APOSOR480596-100GEA
69.6
EUR
APOSOR480596-100G
APOSOR480596-25G
L-Prolinbenzylester -hydrochlorid 98%
L-Prolinbenzylester -hydrochlorid
Formel:
C₁₂H₁₅NO₂ Schmelzpunkt: 148…151 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00039002 CAS-Nummer: 16652-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...