4-Amino-1,2,5-thiadiazole-3-carboxylic acid 97%
Lieferant: Apollo Scientific
OR480213-10G
OR480213-1G
OR480213-5G
APOSOR480213-10GEA
908
EUR
APOSOR480213-10G
APOSOR480213-1G
APOSOR480213-5G
4-Amino-1,2,5-thiadiazole-3-carboxylic acid 97%
4-Amino-1,2,5-thiadiazole-3-carboxylic acid
Formel:
C₃H₃N₃O₂S Schmelzpunkt: 217…218 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00718830 CAS-Nummer: 2829-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...