2-(1-Piperidinyl)-3-pyridinamine 97%
Lieferant: Apollo Scientific
OR471683-1G
OR471683-5G
APOSOR471683-1GEA
364
EUR
APOSOR471683-1G
APOSOR471683-5G
2-(1-Piperidinyl)-3-pyridinamine 97%
2-(1-Piperidinyl)-3-pyridinamine
Formel:
C₁₀H₁₅N₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08700153 CAS-Nummer: 5028-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...