4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98%
Lieferant: Apollo Scientific
Synonyme:
Fmoc-Asp(OBzl)-OH, 4-Benzyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester
OR471606-25G
OR471606-100G
APOSOR471606-25GEA
71.2
EUR
APOSOR471606-25G
APOSOR471606-100G
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98%
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
Formel:
C₂₆H₂₃NO₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00065630 CAS-Nummer: 86060-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...