5-Bromo-1H-pyrazol-3-amine 97%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-5-bromopyrazole
OR471180-1G
OR471180-25G
OR471180-5G
APOSOR471180-1GEA
69.6
EUR
APOSOR471180-1G
APOSOR471180-25G
APOSOR471180-5G
5-Bromo-1H-pyrazol-3-amine 97%
5-Bromo-1H-pyrazol-3-amine
Formel:
C3H4BrN3 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22199259 CAS-Nummer: 950739-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...