3-(tert-Butoxy)benzenecarboxylic acid
Lieferant: Apollo Scientific
OR470339-5G
OR470339-1G
APOSOR470339-5GEA
1120
EUR
APOSOR470339-5G
APOSOR470339-1G
3-(tert-Butoxy)benzenecarboxylic acid
3-(tert-Butoxy)benzenecarboxylic acid
Formel:
C₁₁H₁₄O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03412397 CAS-Nummer: 15360-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...