3-(2-Methylphenoxy)propionsäure

Lieferant: Apollo Scientific

OR470104-1G OR470104-5G
APOSOR470104-1GEA 154 EUR
APOSOR470104-1G APOSOR470104-5G
3-(2-Methylphenoxy)propionsäure
3-(2-Methylphenoxy)propionsäure
Formel: C₁₀H₁₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00642410
CAS-Nummer: 25173-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR