Fmoc-L-(2,6-di-Me)Tyr-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Tyr(2,6-DiMe)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid, 2,6-Dimethyl-L-tyrosine, N-FMOC protected

OR46613-1G OR46613-500MG OR46613-250MG
APOSOR46613-1GEA 142 EUR
APOSOR46613-1G APOSOR46613-500MG APOSOR46613-250MG
Fmoc-L-(2,6-di-Me)Tyr-OH
Fmoc-L-(2,6-di-Me)Tyr-OH
Formel: C₂₆H₂₅NO₅
Molekulargewicht: 431.49 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01632251
CAS-Nummer: 206060-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR