(R,R)-(-)-2,5-Hexandiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,5R)-2,5-Hexanediol, (R,R)-(-)-2,5-Dihydroxy hexane, (2R,5R)-Hexane-2,5-diol
OR46574-1G
OR46574-5G
APOSOR46574-1GEA
68.3
EUR
APOSOR46574-1G
APOSOR46574-5G
(R,R)-(-)-2,5-Hexandiol
(R,R)-(-)-2,5-Hexandiol
Formel:
C₆H₁₄O₂ Schmelzpunkt: 52…54 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00142339 CAS-Nummer: 17299-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...