Palladium(2+)chloridacetonitril 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)
OR46250-1G
OR46250-5G
APOSOR46250-1GEA
87
EUR
APOSOR46250-1G
APOSOR46250-5G
Palladium(2+)chloridacetonitril 95%
Bis(acetonitrile)palladium(II) dichloride
Formel:
C₄H₆Cl₂N₂Pd Schmelzpunkt: 300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013122 CAS-Nummer: 14592-56-4 EINECS: 238-637-3 UN-Nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...