2,3-Dibrompentan

Lieferant: Apollo Scientific

OR460065-1G OR460065-5G
APOSOR460065-1GEA 104 EUR
APOSOR460065-1G APOSOR460065-5G
2,3-Dibrompentan
2,3-Dibromopentane
Formel: H₃C(BrCH)₂CH₂CH₃
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 5398-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR