Ethyl (S)-2-oxiranylacetate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl (S)-3,4-epoxybutanoate

OR4600-500MG OR4600-1G
APOSOR4600-500MGEA 1090 EUR
APOSOR4600-500MG APOSOR4600-1G
Ethyl (S)-2-oxiranylacetate 97%
Ethyl-(S)-2-oxiranylacetate
Formel: C₆H₁₀O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00672889
CAS-Nummer: 112083-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR