3-Amino-1-benzylpyrrolidine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR451185-1G OR451185-250MG
APOSOR451185-1GEA 198 EUR
APOSOR451185-1G APOSOR451185-250MG
3-Amino-1-benzylpyrrolidine-3-carboxylic acid 97%
3-Amino-1-benzylpyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₆N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06410530
CAS-Nummer: 145602-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR