4-(4-Aminophenyl)butanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-Aminophenyl)butyric acid
OR43526-1G
OR43526-5G
APOSOR43526-1GEA
63.8
EUR
APOSOR43526-1G
APOSOR43526-5G
4-(4-Aminophenyl)butanoic acid
4-(4-Aminophenyl)butanoic acid
Formel:
C₁₀H₁₃NO₂ Schmelzpunkt: 121…133 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00075052 CAS-Nummer: 15118-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...