3-(1H-Imidazol-4-yl)propanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Deamino-histidine, Imidazole-5-propionic acid, Imidazolepropionic acid

OR4318-1G OR4318-250MG
APOSOR4318-1GEA 299 EUR
APOSOR4318-1G APOSOR4318-250MG
3-(1H-Imidazol-4-yl)propanoic acid
3-(1H-Imidazol-4-yl)propanoic acid
Formel: C₆H₈N₂O₂
Molekulargewicht: 140.142 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00237415
CAS-Nummer: 1074-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR