3-Cyan-2-(4-morpholino)pyridin
Lieferant: Apollo Scientific
Synonyme:
2-(Morpholin-4-yl)nicotinonitrile
OR40760-1G
OR40760-5G
OR40760-10G
APOSOR40760-1GEA
0
EUR
APOSOR40760-1G
APOSOR40760-5G
APOSOR40760-10G
3-Cyan-2-(4-morpholino)pyridin
3-Cyan-2-(4-morpholino)pyridin
Formel:
C₁₀H₁₁N₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04113612 CAS-Nummer: 59025-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...