5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-5-bromo-1H-pyrazolo[3,4-b]pyridine
OR40403-1G
OR40403-250MG
OR40403-5G
APOSOR40403-1GEA
29
EUR
APOSOR40403-1G
APOSOR40403-250MG
APOSOR40403-5G
5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine
5-Bromo-1H-pyrazolo[3,4-b]pyridin-3-amine
Formel:
C₆H₅BrN₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08059261 CAS-Nummer: 405224-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...