4-Brom-2-pyridon 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine
OR40285-1G
OR40285-5G
APOSOR40285-1GEA
21.9
EUR
APOSOR40285-1G
APOSOR40285-5G
4-Brom-2-pyridon 99%
4-Brom-2-pyridon
Formel:
C₅H₄BrNO Schmelzpunkt: 201…204 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00234041 CAS-Nummer: 36953-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...